Gene Transcripts

Gene Symbol
Uchl4
Entrez Gene ID
93841
Gene Name
ubiquitin carboxyl-terminal esterase L4
Gene Aliases
Chromosome Location
Chr. 9: 64235201 - 64236362 on Build GRCm38
UniGene ID
Mm.390180
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033607.1 NP_291085.1 1 160
GenBank mRNA AB035420.1 - 1 160
BC139440.1 - 1 151 8862462
BC139441.1 - 1 151 8862463

View Full Product Details