Gene Transcripts
- Gene Symbol
- Vmn1r13
- Entrez Gene ID
- 113862
- Gene Name
- vomeronasal 1 receptor 13
- Gene Aliases
- V1rc5
- Chromosome Location
- Chr. 6: 57203298 - 57210760 on Build GRCm38
- UniGene ID
- Mm.431976
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053235.2 |
NP_444465.2
|
1 |
109 |
|
View Full Product Details