Gene Transcripts

Gene Symbol
SSH1
Entrez Gene ID
54434
Gene Name
slingshot protein phosphatase 1
Gene Aliases
SSH1L
Chromosome Location
Chr. 12: 108782690 - 108857606 on Build GRCh38
UniGene ID
Hs.199763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161330.1 NP_001154802.1 7 592
NM_001161331.1 NP_001154803.1 6 684
NM_018984.3 NP_061857.3 7 592
XM_005268984.4 XP_005269041.1 7 1303
XM_005268985.2 XP_005269042.1 6 418
XM_011538497.1 XP_011536799.1 6 684
XM_011538499.1 XP_011536801.1 6 684
XM_011538500.2 XP_011536802.1 6 508
XM_011538501.2 XP_011536803.1 6 504
XM_017019491.1 XP_016874980.1 6 1530
XM_017019492.1 XP_016874981.1 7 645
XM_017019493.1 XP_016874982.1 6 436
XM_017019494.1 XP_016874983.1 6 531
GenBank mRNA AB072355.1 - 7 505
AB072356.1 - 7 505
AB072357.1 - 3 773
AK095421.1 - 6 685
BC062341.1 - 7 584 6376684

View Full Product Details