Gene Transcripts

Gene Symbol
PRSS41
Entrez Gene ID
360226
Gene Name
protease, serine 41
Gene Aliases
TESSP1
Chromosome Location
Chr. 16: 2789525 - 2805287 on Build GRCh38
UniGene ID
Hs.647401
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135086.1 NP_001128558.1 3 385
XM_011522477.2 XP_011520779.1 5 1003

View Full Product Details