Gene Transcripts
- Gene Symbol
- PPA1
- Entrez Gene ID
- 5464
- Gene Name
- pyrophosphatase (inorganic) 1
- Gene Aliases
- HEL-S-66p, IOPPP, PP, PP1, SID6-8061
- Chromosome Location
- Chr. 10: 70202830 - 70233434 on Build GRCh38
- UniGene ID
- Hs.437403
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021129.3 |
NP_066952.1
|
2, 3 |
233 |
|
GenBank mRNA |
AB026723.1 |
- |
2, 3 |
156 |
|
AF092439.1 |
- |
2, 3 |
203 |
|
AF108211.1 |
- |
2, 3 |
114 |
|
AF119665.1 |
- |
2, 3 |
203 |
|
AF154065.1 |
- |
2, 3 |
171 |
|
AF217186.1 |
- |
2, 3 |
126 |
|
AK058193.1 |
- |
2, 3 |
226 |
|
BC001022.2 |
- |
2, 3 |
208 |
3345496 |
BC061581.1 |
- |
2, 3 |
216 |
6526628 |
BC105034.1 |
- |
2, 3 |
199 |
8144037 |
BC105036.1 |
- |
2, 3 |
199 |
8144039 |
BC107882.1 |
- |
2, 3 |
167 |
6495107 |
BU520836.1 |
- |
2, 3 |
214 |
|
EU794626.1 |
- |
2, 3 |
233 |
|
View Full Product Details