Gene Transcripts

Gene Symbol
ITGA4
Entrez Gene ID
3676
Gene Name
integrin subunit alpha 4
Gene Aliases
CD49D, IA4
Chromosome Location
Chr. 2: 181456892 - 181538928 on Build GRCh38
UniGene ID
Hs.440955
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000885.5 NP_000876.3 14 2292
GenBank mRNA AB208885.1 - 11 1070
BC016671.2 - 14 1757
L12002.1 - 14 1957
X16983.1 - 14 1571

View Full Product Details