Gene Transcripts

Gene Symbol
RDH16
Entrez Gene ID
8608
Gene Name
retinol dehydrogenase 16 (all-trans)
Gene Aliases
RODH-4, SDR9C8
Chromosome Location
Chr. 12: 56951431 - 56959374 on Build GRCh38
UniGene ID
Hs.134958
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320108.1 NP_001307037.1 3 3150
NM_003708.4 NP_003699.3 4 3409
GenBank mRNA AB209332.1 - 3 3150
AF086735.1 - 4 2343

View Full Product Details