Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001165412.1 | NP_001158884.1 | 6 | 832 | |
NM_001319226.1 | NP_001306155.1 | 6 | 642 | ||
NM_003998.3 | NP_003989.2 | 6 | 835 | ||
XM_011532006.2 | XP_011530308.1 | 7 | 602 | ||
XM_011532008.2 | XP_011530310.1 | 5 | 375 | ||
GenBank mRNA | AK291450.1 | - | 6 | 496 | |
BC051765.1 | - | 6 | 835 | 6500087 | |
CR749522.1 | - | 6 | 592 | ||
M55643.1 | - | 6 | 549 | ||
M58603.1 | - | 6 | 762 |