Gene Transcripts

Gene Symbol
NFIA
Entrez Gene ID
4774
Gene Name
nuclear factor I A
Gene Aliases
CTF, NF-I/A, NF1-A, NFI-A, NFI-L
Chromosome Location
Chr. 1: 61077124 - 61462788 on Build GRCh38
UniGene ID
Hs.740757
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134673.3 NP_001128145.1 2 907
NM_001145511.1 NP_001138983.1 2 751
NM_001145512.1 NP_001138984.1 3 640
NM_005595.4 NP_005586.1 2 907
XM_011541512.2 XP_011539814.1 2 968
XM_011541514.2 XP_011539816.1 2 901
XM_011541515.2 XP_011539817.1 2 968
XM_017001362.1 XP_016856851.1 2 829
GenBank mRNA AB037860.1 - 2 907
AK299356.1 - 2 648
AK299579.1 - 3 640
BC022264.1 - 2 594 4270934
U07809.1 - 1 370

View Full Product Details