Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304495.1 | NP_001291424.1 | 2 | 563 | |
NM_171982.4 | NP_741983.2 | 3 | 659 | ||
XM_005273452.3 | XP_005273509.1 | 3 | 654 | ||
XM_011544453.1 | XP_011542755.1 | 4 | 687 | ||
GenBank mRNA | AB029021.1 | - | 3 | 609 | |
AF492463.1 | - | 3 | 657 | ||
AK308236.1 | - | 2 | 483 | ||
BC018337.1 | - | 4 | 769 | ||
BC069226.1 | - | 3 | 601 | 6428667 |