Gene Transcripts

Gene Symbol
UNC5D
Entrez Gene ID
137970
Gene Name
unc-5 netrin receptor D
Gene Aliases
PRO34692, Unc5h4
Chromosome Location
Chr. 8: 35235457 - 35796550 on Build GRCh38
UniGene ID
Hs.238889
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322818.1 NP_001309747.1 4 552
NM_080872.3 NP_543148.2 4 839
XM_011544400.2 XP_011542702.1 4 813
XM_011544401.2 XP_011542703.1 4 813
XM_011544402.2 XP_011542704.1 4 813
XM_011544403.2 XP_011542705.1 4 813
XM_011544404.2 XP_011542706.1 4 813
XM_017013044.1 XP_016868533.1 4 814
XM_017013045.1 XP_016868534.1 4 814
XM_017013046.1 XP_016868535.1 4 814
XM_017013047.1 XP_016868536.1 4 813
XM_017013048.1 XP_016868537.1 4 1999
XM_017013049.1 XP_016868538.1 4 1950
XM_017013050.1 XP_016868539.1 5 1406
GenBank mRNA AB055056.1 - 4 551
AY358147.1 - 4 839
BY798317.2 - 4 459

View Full Product Details