Gene Transcripts

Gene Symbol
PASD1
Entrez Gene ID
139135
Gene Name
PAS domain containing 1
Gene Aliases
CT63, OXTES1
Chromosome Location
Chr. X: 151563535 - 151676739 on Build GRCh38
UniGene ID
Hs.160594
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173493.2 NP_775764.2 15, 16 2515
XM_011531102.1 XP_011529404.1 15, 16 2425
GenBank mRNA AK097552.1 - 15, 16 2428
AY270020.1 - 14, 15 3587
AY623425.1 - 3, 4 779
BC040301.1 - 15, 16 2326 4826573

View Full Product Details