Gene Transcripts

Gene Symbol
QSOX2
Entrez Gene ID
169714
Gene Name
quiescin sulfhydryl oxidase 2
Gene Aliases
QSCN6L1, SOXN
Chromosome Location
Chr. 9: 136206333 - 136245841 on Build GRCh38
UniGene ID
Hs.144073
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181701.3 NP_859052.3 11 1498
GenBank mRNA AJ318051.1 - 11 1498
AK128077.1 - 11 1163
AK128750.1 - 5 646
AL834369.1 - 8 989
BC047604.2 - 12 1380

View Full Product Details