Gene Transcripts

Gene Symbol
TSSK1B
Entrez Gene ID
83942
Gene Name
testis specific serine kinase 1B
Gene Aliases
FKSG81, SPOGA4, STK22D, TSK1, TSSK1
Chromosome Location
Chr. 5: 113432554 - 113435031 on Build GRCh38
UniGene ID
Hs.519507
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032028.3 NP_114417.1 1 1241
GenBank mRNA AF348076.1 - 1 1133
AK122904.1 - 1 1199
AK313332.1 - 1 1194
AY028964.1 - 1 1139
BC022515.1 - 1 1179 4816988

View Full Product Details