Gene Transcripts

Gene Symbol
OGFR
Entrez Gene ID
11054
Gene Name
opioid growth factor receptor
Gene Aliases
Chromosome Location
Chr. 20: 62804825 - 62814000 on Build GRCh38
UniGene ID
Hs.67896
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007346.2 NP_031372.2 3 315
GenBank mRNA AF109134.1 - 3 308
AF172449.1 - 3 313
AF172450.1 - 3 313
AF172451.1 - 3 313
AF172452.1 - 3 313
AF172453.1 - 313
AK022234.1 - 3 305
AK024485.1 - 1 2401
AK314696.1 - 3 307
BC008768.2 - 3 290 3050950
BC014137.2 - 3 290 4643904
BC032666.1 - 3 270 5552531
BM551600.1 - 3 315

View Full Product Details