Gene Transcripts

Gene Symbol
QPCT
Entrez Gene ID
25797
Gene Name
glutaminyl-peptide cyclotransferase
Gene Aliases
GCT, QC, sQC
Chromosome Location
Chr. 2: 37344610 - 37373322 on Build GRCh38
UniGene ID
Hs.79033
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012413.3 NP_036545.1 5 927
GenBank mRNA AK290605.1 - 5 909
BC020456.1 - 5 828
BC036721.1 - 5 806 5224222
BC047756.2 - 5 904 6058335
X67731.1 - 4 609
X71125.1 - 5 816

View Full Product Details