Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000170.2 | NP_000161.2 | 1, 2 | 458 | |
GenBank mRNA | AK314156.1 | - | 1, 2 | 460 | |
BC111993.1 | - | 1, 2 | 331 | 8327461 | |
BC111995.1 | - | 1, 2 | 331 | 8327463 | |
D90239.1 | - | 1, 2 | 415 | ||
DR002916.1 | - | 1, 2 | 443 | ||
M63635.1 | - | 1, 2 | 346 | ||
M64590.1 | - | 1, 2 | 415 |