Gene Transcripts

Gene Symbol
LCN1
Entrez Gene ID
3933
Gene Name
lipocalin 1
Gene Aliases
PMFA, TLC, TP, VEGP
Chromosome Location
Chr. 9: 135521296 - 135526540 on Build GRCh38
UniGene ID
Hs.530311
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252617.1 NP_001239546.1 1, 2 146
NM_001252618.1 NP_001239547.1 1, 2 146
NM_001252619.1 NP_001239548.1 1, 2 146
NM_002297.3 NP_002288.1 1, 2 146
XM_011518675.2 XP_011516977.1 1, 2 288
GenBank mRNA BC065721.1 - 1, 2 148
BC074925.2 - 1, 2 110 30915261
BC074926.2 - 1, 2 110 30915486
CD721968.1 - 1, 2 126
CK429694.1 - 1, 2 123
CK430667.1 - 1, 2 134
M90424.1 - 1, 2 146
X62418.1 - 1, 2 108
X67647.1 - 1, 2 129

View Full Product Details