Gene Transcripts

Gene Symbol
DPP8
Entrez Gene ID
54878
Gene Name
dipeptidyl peptidase 8
Gene Aliases
DP8, DPRP-1, DPRP1, MST097, MSTP097, MSTP135, MSTP141
Chromosome Location
Chr. 15: 65442463 - 65517713 on Build GRCh38
UniGene ID
Hs.458609
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320875.1 NP_001307804.1 12 1801
NM_001320876.1 NP_001307805.1 11 1770
NM_017743.5 NP_060213.2 11 1665
NM_130434.4 NP_569118.1 11 1665
NM_197960.3 NP_932064.1 12 1706
NM_197961.3 NP_932065.1 12 1801
NR_135485.1 11 1665
NR_135486.1 12 1706
XM_005254505.1 XP_005254562.1 3 347
XM_011521730.1 XP_011520032.1 10 2925
XM_011521731.1 XP_011520033.1 10 2880
XM_011521732.2 XP_011520034.1 11 3054
XM_011521733.1 XP_011520035.1 10 1472
XM_011521734.1 XP_011520036.1 11 3054
XM_017022373.1 XP_016877862.1 11 3054
XM_017022374.1 XP_016877863.1 11 3054
XM_017022375.1 XP_016877864.1 10 1517
XM_017022376.1 XP_016877865.1 11 1646
XM_017022377.1 XP_016877866.1 9 1343
XM_017022378.1 XP_016877867.1 11 3054
XM_017022379.1 XP_016877868.1 10 1472
XM_017022380.1 XP_016877869.1 10 1472
XM_017022381.1 XP_016877870.1 9 1343
XM_017022382.1 XP_016877871.1 4 474
XM_017022383.1 XP_016877872.1 4 446
GenBank mRNA AF221634.1 - 11 1639
AF221636.1 - 4 476
AK000290.1 - 11 1587
AK027826.1 - 7 844
AK291057.1 - 12 1776
AK296915.1 - 11 1741
AK302638.1 - 12 1642
AY172659.1 - 10 1433
AY354202.1 - 12 1634
BC030688.2 - 12 1792 4822550

View Full Product Details