Gene Transcripts

Gene Symbol
MPDZ
Entrez Gene ID
8777
Gene Name
multiple PDZ domain crumbs cell polarity complex component
Gene Aliases
HYC2, MUPP1
Chromosome Location
Chr. 9: 13105296 - 13279687 on Build GRCh38
UniGene ID
Hs.169378
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001261406.1 NP_001248335.1 4 602
NM_001261407.1 NP_001248336.1 4 602
NM_003829.4 NP_003820.2 4 602
XM_005251622.4 XP_005251679.1 4 551
XM_005251623.4 XP_005251680.1 4 551
XM_006716885.3 XP_006716948.1 4 600
XM_006716886.3 XP_006716949.1 5 759
XM_006716887.3 XP_006716950.1 4 703
XM_006716888.3 XP_006716951.1 4 551
XM_006716889.3 XP_006716952.1 4 554
XM_006716891.3 XP_006716954.1 4 554
XM_017015252.1 XP_016870741.1 4 551
XM_017015253.1 XP_016870742.1 4 539
XM_017015254.1 XP_016870743.1 5 762
XM_017015255.1 XP_016870744.1 4 610
XM_017015256.1 XP_016870745.1 4 551
XM_017015257.1 XP_016870746.1 4 538
XM_017015258.1 XP_016870747.1 4 551
XM_017015259.1 XP_016870748.1 4 551
GenBank mRNA AB210041.1 - 4 603
AF093419.1 - 3 426
BC140793.1 - 3 426 9021661
BC144564.1 - 3 426 9053092

View Full Product Details