Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016089.2 | NP_057173.2 | 4 | 661 | |
XM_017006575.1 | XP_016862064.1 | 4 | 742 | ||
XM_017006576.1 | XP_016862065.1 | 6 | 891 | ||
XM_017006577.1 | XP_016862066.1 | 4 | 1947 | ||
XM_017006578.1 | XP_016862067.1 | 4 | 722 | ||
XM_017006579.1 | XP_016862068.1 | 6 | 847 | ||
GenBank mRNA | AB209387.1 | - | 4 | 642 | |
AF114816.1 | - | 6 | 964 | ||
AF114817.1 | - | 6 | 964 | ||
AL832831.1 | - | 4 | 646 | ||
AY258146.1 | - | 4 | 661 | ||
BC005859.2 | - | 4 | 620 | ||
BC006247.2 | - | 4 | 641 | ||
BC048798.1 | - | 4 | 638 |