Gene Transcripts

Gene Symbol
NDUFB5
Entrez Gene ID
4711
Gene Name
NADH:ubiquinone oxidoreductase subunit B5
Gene Aliases
CISGDH, SGDH
Chromosome Location
Chr. 3: 179604787 - 179624500 on Build GRCh38
UniGene ID
Hs.730674
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199957.1 NP_001186886.1 3, 4 319
NM_002492.3 NP_002483.1 5, 6 475
GenBank mRNA AF047181.1 - 5, 6 452
AK313822.1 - 5, 6 453
BC005271.1 - 5, 6 466 3997377
BC009796.1 - 5, 6 470 4284776
BC017909.1 - 5, 6 465
BC093071.1 - 3, 4 296 30325282
BG776403.1 - 5, 6 482
BX648034.1 - 3, 4 313
DA112751.1 - 5, 6 478

View Full Product Details