Gene Transcripts

Gene Symbol
SRM
Entrez Gene ID
6723
Gene Name
spermidine synthase
Gene Aliases
PAPT, SPDSY, SPS1, SRML1
Chromosome Location
Chr. 1: 11054592 - 11060053 on Build GRCh38
UniGene ID
Hs.76244
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003132.2 NP_003123.2 8 1114
GenBank mRNA BC000309.2 - 8 1080
BC033106.1 - 8 1046 5420683
M34338.1 - 8 1105

View Full Product Details