Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014228.4 | NP_055043.2 | 7 | 1416 | |
XM_017009767.1 | XP_016865256.1 | 8 | 1323 | ||
XM_017009768.1 | XP_016865257.1 | 8 | 1399 | ||
XM_017009769.1 | XP_016865258.1 | 8 | 2930 | ||
XM_017009770.1 | XP_016865259.1 | 8 | 1050 | ||
GenBank mRNA | AK096607.1 | - | 7 | 1320 | |
AK308895.1 | - | 7 | 1192 | ||
AK312309.1 | - | 7 | 1196 | ||
BC069631.1 | - | 7 | 949 | 7262437 | |
BC093785.1 | - | 7 | 1025 | 7939630 | |
BC113425.1 | - | 7 | 1025 | 8322477 | |
S80071.1 | - | 7 | 949 |