Gene Transcripts
- Gene Symbol
- SLC5A11
- Entrez Gene ID
- 115584
- Gene Name
- solute carrier family 5 member 11
- Gene Aliases
- KST1, RKST1, SGLT6, SMIT2
- Chromosome Location
- Chr. 16: 24832784 - 24911711 on Build GRCh38
- UniGene ID
- Hs.164118
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001258411.2 |
NP_001245340.1
|
5, 6 |
687 |
|
NM_001258413.2 |
NP_001245342.1
|
4, 5 |
528 |
|
NM_001258414.1 |
NP_001245343.1
|
4, 5 |
528 |
|
NM_052944.4 |
NP_443176.2
|
4, 5 |
538 |
|
XM_005255080.2 |
XP_005255137.1
|
5, 6 |
566 |
|
XM_006721015.2 |
XP_006721078.1
|
4, 5 |
651 |
|
XM_011545725.2 |
XP_011544027.2
|
5, 6 |
814 |
|
XM_017022900.1 |
XP_016878389.1
|
5, 6 |
882 |
|
XM_017022901.1 |
XP_016878390.1
|
5, 6 |
1592 |
|
XM_017022902.1 |
XP_016878391.1
|
5, 6 |
721 |
|
XM_017022903.1 |
XP_016878392.1
|
6, 7 |
731 |
|
XM_017022904.1 |
XP_016878393.1
|
6, 7 |
901 |
|
XM_017022905.1 |
XP_016878394.1
|
5, 6 |
882 |
|
XM_017022906.1 |
XP_016878395.1
|
5, 6 |
742 |
|
XM_017022907.1 |
XP_016878396.1
|
5, 6 |
566 |
|
XM_017022908.1 |
XP_016878397.1
|
5, 6 |
625 |
|
XM_017022909.1 |
XP_016878398.1
|
5, 6 |
566 |
|
GenBank mRNA |
AF292385.1 |
- |
3, 4 |
306 |
|
AJ305237.1 |
- |
3, 4 |
324 |
|
AK125267.1 |
- |
6, 7 |
654 |
|
AK298373.1 |
- |
3, 4 |
323 |
|
AK303691.1 |
- |
5, 6 |
555 |
|
AY044906.1 |
- |
4, 5 |
538 |
|
BC049385.1 |
- |
4, 5 |
398 |
5755223 |
BC057780.1 |
- |
4, 5 |
397 |
5287366 |
U41898.1 |
- |
1, 2 |
90 |
|
View Full Product Details