Gene Transcripts

Gene Symbol
NARS
Entrez Gene ID
4677
Gene Name
asparaginyl-tRNA synthetase
Gene Aliases
ASNRS, NARS1
Chromosome Location
Chr. 18: 57600656 - 57621945 on Build GRCh38
UniGene ID
Hs.465224
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004539.3 NP_004530.1 3, 4 488
XM_005266700.1 XP_005266757.1 3, 4 266
GenBank mRNA AJ000334.1 - 3, 4 333
AK129897.1 - 3, 4 291
AK222835.1 - 3, 4 335
AK293985.1 - 2, 3 268
AK294364.1 - 3, 4 335
AK294763.1 - 3, 4 319
AK297777.1 - 3, 4 328
BC001687.1 - 3, 4 337 2987840
D84273.1 - 3, 4 292
DC302277.1 - 3, 4 489

View Full Product Details