Gene Transcripts

Gene Symbol
CDK2
Entrez Gene ID
1017
Gene Name
cyclin dependent kinase 2
Gene Aliases
CDKN2, p33(CDK2)
Chromosome Location
Chr. 12: 55966769 - 55972789 on Build GRCh38
UniGene ID
Hs.19192
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290230.1 NP_001277159.1 5 1666
NM_001798.4 NP_001789.2 7 1846
NM_052827.3 NP_439892.2 6 1744
XM_011537732.1 XP_011536034.1 8 1990
GenBank mRNA BC003065.2 - 7 1766
M68520.1 - 7 1795

View Full Product Details