Gene Transcripts

Gene Symbol
MID2
Entrez Gene ID
11043
Gene Name
midline 2
Gene Aliases
FXY2, MRX101, RNF60, TRIM1
Chromosome Location
Chr. X: 107820961 - 107931637 on Build GRCh38
UniGene ID
Hs.12256
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012216.3 NP_036348.2 3 1324
NM_052817.2 NP_438112.2 3 1324
XM_005262062.4 XP_005262119.1 3 5587
XM_017029239.1 XP_016884728.1 3 5587
GenBank mRNA AF196481.1 - 3 764
AY625004.1 - 2 691
BC017707.1 - 2 718 4446324
BT006663.1 - 2 691
Y18880.1 - 3 948

View Full Product Details