Gene Transcripts
- Gene Symbol
- VNN2
- Entrez Gene ID
- 8875
- Gene Name
- vanin 2
- Gene Aliases
- FOAP-4, GPI-80
- Chromosome Location
- Chr. 6: 132743503 - 132763489 on Build GRCh38
- UniGene ID
- Hs.293130
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004665.4 |
NP_004656.2
|
4, 5 |
945 |
|
NM_078488.2 |
NP_511043.1
|
5, 6 |
773 |
|
XM_006715593.3 |
XP_006715656.1
|
6, 7 |
920 |
|
XM_011536231.1 |
XP_011534533.1
|
4, 5 |
841 |
|
XM_017011407.1 |
XP_016866896.1
|
5, 6 |
1556 |
|
XM_017011408.1 |
XP_016866897.1
|
6, 7 |
925 |
|
XM_017011409.1 |
XP_016866898.1
|
6, 7 |
818 |
|
GenBank mRNA |
AB026705.1 |
- |
4, 5 |
827 |
|
AJ132100.1 |
- |
4, 5 |
831 |
|
AK290908.1 |
- |
4, 5 |
841 |
|
AK292125.1 |
- |
5, 6 |
761 |
|
BC126145.1 |
- |
4, 5 |
907 |
8991861 |
BC126147.1 |
- |
4, 5 |
907 |
8991863 |
D89974.1 |
- |
4, 5 |
831 |
|
View Full Product Details