Gene Transcripts

Gene Symbol
ADCY4
Entrez Gene ID
196883
Gene Name
adenylate cyclase 4
Gene Aliases
AC4
Chromosome Location
Chr. 14: 24318349 - 24335071 on Build GRCh38
UniGene ID
Hs.443428
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198568.1 NP_001185497.1 8 1545
NM_001198592.1 NP_001185521.1 9 1244
NM_139247.3 NP_640340.2 9 1240
GenBank mRNA AF497516.1 - 8 1132
AK122714.1 - 9 1240
AK126468.1 - 7 966
AK131333.1 - 1 658
BC117473.1 - 8 1221 40126024
BC117475.1 - 8 1221 40126026
BC143947.1 - 8 1253
BX248285.1 - 9 1211
BX537489.1 - 2 2456

View Full Product Details