Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146008.1 | NP_001139480.1 | 1 | 1240 | |
NM_001146009.1 | NP_001139481.1 | 1 | 1019 | ||
NM_030958.2 | NP_112220.2 | 2 | 1230 | ||
XM_005251313.2 | XP_005251370.1 | 2 | 1230 | ||
XM_017013883.1 | XP_016869372.1 | 2 | 1230 | ||
XM_017013884.1 | XP_016869373.1 | 2 | 1230 | ||
XM_017013886.1 | XP_016869375.1 | 2 | 1230 | ||
GenBank mRNA | AF205075.1 | - | 2 | 1139 | |
AK022760.1 | - | 1 | 1240 | ||
AK096879.1 | - | 2 | 1230 | ||
AK299048.1 | - | 2 | 1186 | ||
AK307567.1 | - | 2 | 1188 | ||
AK308488.1 | - | 2 | 1132 | ||
BC137424.1 | - | 1 | 1025 | 9021422 | |
BC139755.1 | - | 1 | 565 | 40118841 | |
BC144618.1 | - | 1 | 1025 |