Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005001.4 | NP_004992.2 | 4 | 493 | |
GenBank mRNA | AF050637.1 | - | 4 | 450 | |
AF054178.1 | - | 4 | 464 | ||
BC003102.1 | - | 4 | 476 | 3537231 | |
BC107892.1 | - | 4 | 464 | ||
CD172691.1 | - | 4 | 523 | ||
DA189271.1 | - | 4 | 471 |