Gene Transcripts

Gene Symbol
PRRG1
Entrez Gene ID
5638
Gene Name
proline rich and Gla domain 1
Gene Aliases
PRGP1
Chromosome Location
Chr. X: 37349275 - 37457295 on Build GRCh38
UniGene ID
Hs.190341
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000950.2 NP_000941.1 5 2597
NM_001142395.1 NP_001135867.1 4 2506
NM_001173489.1 NP_001166960.1 5 2616
NM_001173490.1 NP_001166961.1 4 2525
GenBank mRNA AF009242.1 - 5 2509
BC030786.2 - 4 2489
BC060833.1 - 4 2445 30348133

View Full Product Details