Gene Transcripts
- Gene Symbol
- WDR46
- Entrez Gene ID
- 9277
- Gene Name
- WD repeat domain 46
- Gene Aliases
- BING4, C6orf11, FP221, UTP7
- Chromosome Location
- Chr. 6: 33278850 - 33289527 on Build GRCh38
- UniGene ID
- Hs.520063
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164267.1 |
NP_001157739.1
|
3, 4 |
563 |
|
NM_005452.5 |
NP_005443.3
|
3, 4 |
725 |
|
XM_017011484.1 |
XP_016866973.1
|
3, 4 |
439 |
|
XM_017011485.1 |
XP_016866974.1
|
1, 2 |
87 |
|
XM_017011487.1 |
XP_016866976.1
|
1, 2 |
87 |
|
GenBank mRNA |
AF447870.1 |
- |
3, 4 |
417 |
|
AK292171.1 |
- |
3, 4 |
708 |
|
AK292616.1 |
- |
3, 4 |
508 |
|
AK297925.1 |
- |
3, 4 |
320 |
|
AK298151.1 |
- |
3, 4 |
274 |
|
AK308534.1 |
- |
2, 3 |
201 |
|
AK311388.1 |
- |
3, 4 |
436 |
|
AK315616.1 |
- |
3, 4 |
437 |
|
BC000388.2 |
- |
3, 4 |
412 |
2821198 |
BX439421.2 |
- |
3, 4 |
412 |
|
DB272479.1 |
- |
3, 4 |
436 |
|
View Full Product Details