Gene Transcripts
- Gene Symbol
- UBXN1
- Entrez Gene ID
- 51035
- Gene Name
- UBX domain protein 1
- Gene Aliases
- 2B28, SAKS1, UBXD10
- Chromosome Location
- Chr. 11: 62676498 - 62679117 on Build GRCh38
- UniGene ID
- Hs.351296
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286077.1 |
NP_001273006.1
|
6, 7 |
728 |
|
NM_001286078.1 |
NP_001273007.1
|
4, 5 |
551 |
|
NM_015853.4 |
NP_056937.2
|
6, 7 |
728 |
|
XM_005274033.4 |
XP_005274090.1
|
7, 8 |
641 |
|
XM_017017874.1 |
XP_016873363.1
|
7, 8 |
637 |
|
GenBank mRNA |
AK303471.1 |
- |
5, 6 |
587 |
|
BC000902.2 |
- |
6, 7 |
667 |
2900493 |
BC001372.2 |
- |
6, 7 |
666 |
3051213 |
BC040129.1 |
- |
6, 7 |
703 |
6065505 |
BI855821.1 |
- |
4, 5 |
494 |
|
M68864.1 |
- |
6, 7 |
672 |
|
View Full Product Details