Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_139246.4 | NP_640339.4 | 3 | 855 | |
XM_011518312.2 | XP_011516614.1 | 2 | 318 | ||
XM_017014342.1 | XP_016869831.1 | 3 | 501 | ||
GenBank mRNA | AF258575.1 | - | 2 | 301 | |
AK094043.1 | - | 3 | 568 | ||
AK122809.1 | - | 1 | 969 | ||
AK128615.1 | - | 4 | 521 | ||
AK291199.1 | - | 3 | 476 | ||
BC035604.1 | - | 3 | 502 | 4385905 | |
BX538308.1 | - | 3 | 496 | ||
BX648493.1 | - | 3 | 494 | ||
KU178877.1 | - | 2 | 286 | ||
KU178878.1 | - | 2 | 258 |