Gene Transcripts

Gene Symbol
TPSD1
Entrez Gene ID
23430
Gene Name
tryptase delta 1
Gene Aliases
MCP7-LIKE, MCP7L1, MMCP-7L
Chromosome Location
Chr. 16: 1256272 - 1258493 on Build GRCh38
UniGene ID
Hs.677789
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012217.2 NP_036349.1 4 585
GenBank mRNA AF206664.1 - 3 422
BC069143.1 - 4 576 7217003

View Full Product Details