Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282143.1 | NP_001269072.1 | 5 | 546 | |
NM_001282144.1 | NP_001269073.1 | 5 | 954 | ||
NM_001282358.1 | NP_001269287.1 | 5 | 478 | ||
NM_024618.3 | NP_078894.2 | 5 | 573 | ||
XM_005271669.1 | XP_005271726.1 | 5 | 797 | ||
XM_005271670.1 | XP_005271727.1 | 4 | 450 | ||
XM_005271671.1 | XP_005271728.1 | 4 | 624 | ||
XM_005271672.1 | XP_005271729.1 | 4 | 447 | ||
XM_006718904.1 | XP_006718967.1 | 5 | 831 | ||
XM_011542980.1 | XP_011541282.1 | 5 | 461 | ||
XM_017018294.1 | XP_016873783.1 | 5 | 831 | ||
GenBank mRNA | AB094095.1 | - | 5 | 557 | |
AK056454.1 | - | 4 | 271 | ||
AK291716.1 | - | 5 | 540 | ||
AK303002.1 | - | 4 | 447 | ||
AY245437.1 | - | 4 | 316 | ||
BC110890.1 | - | 5 | 919 | 4937963 | |
BX647705.1 | - | 5 | 531 | ||
DA441499.1 | - | 5 | 478 | ||
EF452237.1 | - | 4 | 316 |