Gene Transcripts

Gene Symbol
C2orf47
Entrez Gene ID
79568
Gene Name
chromosome 2 open reading frame 47
Gene Aliases
Chromosome Location
Chr. 2: 199955317 - 199964125 on Build GRCh38
UniGene ID
Hs.154494
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024520.2 NP_078796.2 6 1224
XM_005246852.2 XP_005246909.1 4 987
GenBank mRNA AK026208.1 - 6 1224
AL832964.1 - 5 543
BC017959.1 - 5 908 4607945

View Full Product Details