Gene Transcripts
- Gene Symbol
- SMARCC2
- Entrez Gene ID
- 6601
- Gene Name
- SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 2
- Gene Aliases
- BAF170, CRACC2, Rsc8
- Chromosome Location
- Chr. 12: 56161852 - 56189567 on Build GRCh38
- UniGene ID
- Hs.236030
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130420.1 |
NP_001123892.1
|
8, 9 |
822 |
|
NM_003075.3 |
NP_003066.2
|
8, 9 |
822 |
|
NM_139067.2 |
NP_620706.1
|
8, 9 |
822 |
|
XM_005269101.1 |
XP_005269158.1
|
8, 9 |
759 |
|
XM_005269102.1 |
XP_005269159.1
|
8, 9 |
759 |
|
XM_005269103.1 |
XP_005269160.1
|
8, 9 |
759 |
|
XM_005269104.1 |
XP_005269161.1
|
8, 9 |
759 |
|
XM_011538693.2 |
XP_011536995.1
|
7, 8 |
668 |
|
XM_017019884.1 |
XP_016875373.1
|
8, 9 |
759 |
|
XM_017019885.1 |
XP_016875374.1
|
8, 9 |
759 |
|
XM_017019886.1 |
XP_016875375.1
|
8, 9 |
759 |
|
XM_017019887.1 |
XP_016875376.1
|
7, 8 |
646 |
|
GenBank mRNA |
AB209006.1 |
- |
8, 9 |
822 |
|
AB209293.1 |
- |
8, 9 |
731 |
|
AK293896.1 |
- |
8, 9 |
709 |
|
AK315153.1 |
- |
8, 9 |
722 |
|
BC013045.1 |
- |
8, 9 |
748 |
4180388 |
BC026222.2 |
- |
8, 9 |
741 |
4395012 |
BT009924.1 |
- |
8, 9 |
716 |
|
U66616.1 |
- |
8, 9 |
738 |
|
View Full Product Details