Gene Transcripts
- Gene Symbol
- MBOAT2
- Entrez Gene ID
- 129642
- Gene Name
- membrane bound O-acyltransferase domain containing 2
- Gene Aliases
- LPAAT, LPCAT4, LPEAT, LPLAT 2, OACT2
- Chromosome Location
- Chr. 2: 8852690 - 9003750 on Build GRCh38
- UniGene ID
- Hs.467634
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138799.3 |
NP_620154.2
|
1, 2 |
216 |
|
NR_135598.1 |
|
1, 2 |
216 |
|
NR_135599.1 |
|
1, 2 |
216 |
|
NR_135602.1 |
|
1, 2 |
216 |
|
NR_135604.1 |
|
1, 2 |
216 |
|
NR_135606.1 |
|
1, 2 |
216 |
|
XM_011510313.1 |
XP_011508615.1
|
1, 2 |
176 |
|
XM_011510314.2 |
XP_011508616.1
|
1, 2 |
176 |
|
GenBank mRNA |
AB305044.1 |
- |
1, 2 |
80 |
|
AK027321.2 |
- |
1, 2 |
176 |
|
AK295907.1 |
- |
1, 2 |
124 |
|
BC146871.1 |
- |
1, 2 |
160 |
9056799 |
BC157827.1 |
- |
1, 2 |
160 |
9057046 |
BX442603.2 |
- |
1, 2 |
216 |
|
DA236556.1 |
- |
1, 2 |
129 |
|
View Full Product Details