Gene Transcripts
- Gene Symbol
- RGS22
- Entrez Gene ID
- 26166
- Gene Name
- regulator of G-protein signaling 22
- Gene Aliases
- CT145, PRTD-NY2
- Chromosome Location
- Chr. 8: 99960936 - 100106116 on Build GRCh38
- UniGene ID
- Hs.120021
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286692.1 |
NP_001273621.1
|
14, 15 |
2324 |
|
NM_001286693.1 |
NP_001273622.1
|
12, 13 |
2219 |
|
NM_015668.4 |
NP_056483.3
|
14, 15 |
2360 |
|
XM_005250856.2 |
XP_005250913.1
|
14, 15 |
2206 |
|
XM_005250857.2 |
XP_005250914.1
|
14, 15 |
2206 |
|
XM_005250858.2 |
XP_005250915.1
|
14, 15 |
2206 |
|
XM_005250860.2 |
XP_005250917.1
|
14, 15 |
2206 |
|
XM_006716540.2 |
XP_006716603.1
|
14, 15 |
2474 |
|
XM_011516957.1 |
XP_011515259.1
|
15, 16 |
2655 |
|
XM_011516958.1 |
XP_011515260.1
|
14, 15 |
2553 |
|
XM_017013309.1 |
XP_016868798.1
|
14, 15 |
2206 |
|
XM_017013310.1 |
XP_016868799.1
|
14, 15 |
2206 |
|
XM_017013311.1 |
XP_016868800.1
|
7, 8 |
903 |
|
GenBank mRNA |
AK097399.1 |
- |
3, 4 |
393 |
|
AK292559.1 |
- |
14, 15 |
2170 |
|
AK302475.1 |
- |
11, 12 |
1980 |
|
AY009106.1 |
- |
14, 15 |
2153 |
|
BC036665.1 |
- |
14, 15 |
2211 |
5268331 |
BC047060.1 |
- |
15, 16 |
2334 |
|
BC092411.1 |
- |
14, 15 |
2357 |
30386231 |
View Full Product Details