Gene Transcripts

Gene Symbol
NUB1
Entrez Gene ID
51667
Gene Name
negative regulator of ubiquitin like proteins 1
Gene Aliases
BS4, NUB1L, NYREN18
Chromosome Location
Chr. 7: 151341699 - 151378461 on Build GRCh38
UniGene ID
Hs.647082
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243351.1 NP_001230280.1 5, 6 498
NM_016118.4 NP_057202.3 5, 6 498
XM_005250011.2 XP_005250068.1 5, 6 563
XM_017012304.1 XP_016867793.1 5, 6 560
XM_017012305.1 XP_016867794.1 5, 6 560
XM_017012306.1 XP_016867795.1 5, 6 579
XM_017012307.1 XP_016867796.1 5, 6 560
XM_017012308.1 XP_016867797.1 5, 6 560
XM_017012309.1 XP_016867798.1 5, 6 563
XM_017012310.1 XP_016867799.1 6, 7 660
XM_017012311.1 XP_016867800.1 6, 7 564
XM_017012312.1 XP_016867801.1 6, 7 536
XM_017012313.1 XP_016867802.1 6, 7 660
XM_017012314.1 XP_016867803.1 6, 7 536
GenBank mRNA AF155099.1 - 5, 6 430
AF300717.1 - 5, 6 487
AF459743.1 - 5, 6 446
AY129295.1 - 5, 6 446
BC046354.2 - 4, 5 414 6070128
DB048887.1 - 5, 6 498

View Full Product Details