Gene Transcripts

Gene Symbol
STK26
Entrez Gene ID
51765
Gene Name
serine/threonine protein kinase 26
Gene Aliases
MASK, MST4
Chromosome Location
Chr. X: 132023217 - 132075943 on Build GRCh38
UniGene ID
Hs.444247
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042452.1 NP_001035917.1 2 150
NM_016542.3 NP_057626.2 3 441
XM_011531349.1 XP_011529651.1 4 414
XM_011531350.1 XP_011529652.1 3 356
GenBank mRNA AB040057.2 - 3 360
AF231012.1 - 3 329
AF344882.1 - 2 150
AF344883.1 - 2 150
AK075150.1 - 3 356
AK303583.1 - 3 356
AK314356.1 - 3 357
AK316137.1 - 3 356
BC098315.1 - 3 346 40026349
BC099843.1 - 3 346 40026348
BC103503.1 - 3 346 40026350
BT020099.1 - 2 140

View Full Product Details