Gene Transcripts

Gene Symbol
PLXNA3
Entrez Gene ID
55558
Gene Name
plexin A3
Gene Aliases
6.3, HSSEXGENE, PLXN3, PLXN4, XAP-6
Chromosome Location
Chr. X: 154458281 - 154473646 on Build GRCh38
UniGene ID
Hs.632839
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017514.4 NP_059984.3 24 4445
XM_005274705.4 XP_005274762.2 24 4385
XM_005274706.4 XP_005274763.2 24 4117
XM_006724829.3 XP_006724892.1 24 4349
XM_011531183.2 XP_011529485.1 23 3809
GenBank mRNA X87852.1 - 24 4454

View Full Product Details