Gene Transcripts

Gene Symbol
POC5
Entrez Gene ID
134359
Gene Name
POC5 centriolar protein
Gene Aliases
C5orf37
Chromosome Location
Chr. 5: 75674198 - 75717488 on Build GRCh38
UniGene ID
Hs.432726
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099271.1 NP_001092741.1 5 602
NM_152408.2 NP_689621.2 4 401
XM_005248436.1 XP_005248493.1 5 602
XM_011543158.2 XP_011541460.1 5 602
XM_011543159.1 XP_011541461.1 6 608
XM_011543160.1 XP_011541462.1 4 462
XM_017009037.1 XP_016864526.1 5 602
XM_017009038.1 XP_016864527.1 4 522
XM_017009039.1 XP_016864528.1 4 441
XM_017009040.1 XP_016864529.1 4 405
GenBank mRNA AK093098.1 - 4 371
AK296071.1 - 5 519
BC065750.1 - 5 548
BC101325.1 - 4 367 40024984
BC101326.2 - 4 371 40024987
BC101327.2 - 4 371 40024989
BC101328.2 - 4 367
DB448626.1 - 4 401

View Full Product Details