Gene Transcripts

Gene Symbol
WDFY2
Entrez Gene ID
115825
Gene Name
WD repeat and FYVE domain containing 2
Gene Aliases
PROF, WDF2, ZFYVE22
Chromosome Location
Chr. 13: 51584194 - 51766799 on Build GRCh38
UniGene ID
Hs.208550
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052950.3 NP_443182.1 6 848
XM_011534914.1 XP_011533216.1 6 739
XM_011534915.2 XP_011533217.1 5 928
XM_017020386.1 XP_016875875.1 6 6652
GenBank mRNA AF411978.1 - 6 508
AK058149.1 - 4 294
AK309815.1 - 6 701
AK311177.1 - 4 506
BC014004.2 - 6 688 3842589

View Full Product Details