Gene Transcripts

Gene Symbol
FAM26E
Entrez Gene ID
254228
Gene Name
family with sequence similarity 26 member E
Gene Aliases
C6orf188, dJ493F7.3
Chromosome Location
Chr. 6: 116511645 - 116524792 on Build GRCh38
UniGene ID
Hs.105081
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153711.3 NP_714922.1 2 650
GenBank mRNA AK094140.1 - 2 625
AK315572.1 - 2 622
BC032556.1 - 2 673

View Full Product Details