Gene Transcripts

Gene Symbol
EYS
Entrez Gene ID
346007
Gene Name
eyes shut homolog (Drosophila)
Gene Aliases
C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, RP25, SPAM, bA166P24.2, bA307F22.3, bA74E24.1, dJ1018A4.2, dJ22I17.2, dJ303F19.1
Chromosome Location
Chr. 6: 63719980 - 65707225 on Build GRCh38
UniGene ID
Hs.25067
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142800.1 NP_001136272.1 9 1861
NM_001142801.1 NP_001136273.1 9 1861
NM_001292009.1 NP_001278938.1 9 1861
NM_198283.1 NP_938024.1 8 1720
GenBank mRNA AB593114.1 - 9 1826
AY358133.1 - 8 1720
BC133011.1 - 7 1585
BC133013.1 - 7 1582
BX537914.1 - 9 1861
FJ416331.1 - 9 1869
FM209056.1 - 9 1813

View Full Product Details