Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024674.5 | NP_078950.1 | 3 | 419 | |
XM_011542148.1 | XP_011540450.1 | 3 | 409 | ||
GenBank mRNA | AF521099.1 | - | 3 | 370 | |
AK022519.1 | - | 3 | 370 | ||
BC028566.2 | - | 3 | 409 | 3841184 | |
CN349325.1 | - | 3 | 421 |